Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025572.1 | NP_001020743.1 | 8 - 9 | 1154 | 126 | |
GenBank mRNA | AK134530.1 | - | 3 - 4 | 333 | 126 | |
AK153850.1 | - | 7 - 8 | 1063 | 126 | ||
BC023046.1 | - | 8 - 9 | 1025 | 126 | ||
BC050185.1 | - | 8 - 9 | 1134 | 126 | ||
BC064777.1 | - | 7 - 8 | 1069 | 126 | ||
BC080825.1 | - | 7 - 8 | 1077 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |