Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081287.2 | NP_001074756.2 | 4 - 5 | 474 | 83 | |
NM_001161620.1 | NP_001155092.1 | 4 - 5 | 474 | 83 | ||
GenBank mRNA | AK012883.1 | - | 4 - 5 | 449 | 83 | |
AK017344.1 | - | 7 - 8 | 836 | 83 | ||
AK033081.1 | - | 4 - 5 | 434 | 83 | ||
AK036956.1 | - | 4 - 5 | 445 | 83 | ||
AK050710.1 | - | 4 - 5 | 471 | 83 | ||
AK078849.1 | - | 4 - 5 | 473 | 83 | ||
BC049662.1 | - | 4 - 5 | 428 | 83 | ||
BC117550.1 | - | 3 - 4 | 321 | 40105182 | 83 | |
BC118058.1 | - | 3 - 4 | 321 | 40105180 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |