Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163759.1 | NP_001157231.1 | 9 - 10 | 2462 | 93 | |
NM_198942.2 | NP_945180.2 | 8 - 9 | 2303 | 93 | ||
GenBank mRNA | AK053628.1 | - | 9 - 10 | 2130 | 93 | |
BC062952.1 | - | 8 - 9 | 2303 | 6853031 | 93 | |
BC065169.1 | - | 9 - 10 | 2129 | 6831314 | 93 | |
BC066091.1 | - | 8 - 9 | 1965 | 6841207 | 93 | |
BC079618.1 | - | 8 - 9 | 1242 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |