Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pclo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110796.2 | NP_001104266.1 | 15 - 16 | 14305 | 64 | |
NM_011995.4 | NP_036125.4 | 15 - 16 | 14305 | 64 | ||
GenBank mRNA | AB083478.1 | - | 15 - 16 | 14548 | 64 | |
AB093247.1 | - | 10 - 11 | 4190 | 64 | ||
AF181269.2 | - | 4 - 5 | 408 | 64 | ||
Y19185.2 | - | 16 - 17 | 14215 | 64 | ||
Y19186.2 | - | 16 - 17 | 14215 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |