Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026719.2 | NP_080995.2 | 11 - 12 | 1291 | 124 | |
GenBank mRNA | AK032560.1 | - | 1590 | 124 | ||
AK158982.1 | - | 11 - 12 | 1127 | 124 | ||
AK167080.1 | - | 11 - 12 | 1110 | 124 | ||
AY061989.1 | - | 10 - 11 | 1002 | 124 | ||
BC025579.1 | - | 11 - 12 | 1123 | 5325113 | 124 | |
BC027774.1 | - | 11 - 12 | 1174 | 5346846 | 124 | |
BC031902.1 | - | 11 - 12 | 1114 | 5362166 | 124 | |
BC039053.1 | - | 11 - 12 | 1205 | 2655321 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |