Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009592.1 | NP_033722.1 | 9 - 10 | 1227 | 70 | |
GenBank mRNA | AK040069.1 | - | 9 - 10 | 1224 | 70 | |
AK084853.1 | - | 9 - 10 | 1218 | 70 | ||
AK151967.1 | - | 9 - 10 | 1213 | 70 | ||
AK152535.1 | - | 9 - 10 | 1213 | 70 | ||
AK152816.1 | - | 9 - 10 | 1213 | 70 | ||
BC035534.1 | - | 6 - 7 | 856 | 70 | ||
BC151037.1 | - | 9 - 10 | 1227 | 9087950 | 70 | |
U43892.1 | - | 8 - 9 | 1031 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |