Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173019.5 | NP_766607.3 | 2 - 3 | 324 | 60 | |
GenBank mRNA | AK155172.1 | - | 2 - 3 | 262 | 60 | |
AY646092.2 | - | 2 - 3 | 210 | 60 | ||
AY707862.1 | - | 2 - 3 | 227 | 60 | ||
AY756065.1 | - | 2 - 3 | 210 | 60 | ||
AY756066.1 | - | 2 - 3 | 210 | 60 | ||
AY756067.1 | - | 2 - 3 | 210 | 60 | ||
BC057594.1 | - | 2 - 3 | 276 | 6810667 | 60 | |
CA316033.1 | - | 2 - 3 | 276 | 60 | ||
DQ350638.1 | - | 3 - 4 | 296 | 60 | ||
EU571215.1 | - | 2 - 3 | 324 | 60 | ||
EU571216.1 | - | 4 - 5 | 483 | 60 | ||
EU571217.1 | - | 2 - 3 | 323 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |