Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011830.3 | NP_035960.2 | 7 - 8 | 851 | 106 | |
GenBank mRNA | AK011777.1 | - | 7 - 8 | 879 | 106 | |
AK143505.1 | - | 7 - 8 | 892 | 106 | ||
AK146362.1 | - | 7 - 8 | 917 | 106 | ||
AK150713.1 | - | 7 - 8 | 887 | 106 | ||
AK151233.1 | - | 7 - 8 | 887 | 106 | ||
AK151706.1 | - | 7 - 8 | 910 | 106 | ||
AK152691.1 | - | 7 - 8 | 884 | 106 | ||
AK159822.1 | - | 7 - 8 | 884 | 106 | ||
AK160569.1 | - | 7 - 8 | 884 | 106 | ||
AK166667.1 | - | 7 - 8 | 886 | 106 | ||
AK169129.1 | - | 7 - 8 | 917 | 106 | ||
AK169788.1 | - | 7 - 8 | 917 | 106 | ||
BC010314.1 | - | 7 - 8 | 851 | 2646679 | 106 | |
BC052671.1 | - | 7 - 8 | 872 | 30058399 | 106 | |
M33934.1 | - | 7 - 8 | 884 | 106 | ||
M98333.1 | - | 7 - 8 | 818 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |