Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011163.4 | NP_035293.1 | 8 - 9 | 843 | 146 | |
GenBank mRNA | AK028602.1 | - | 8 - 9 | 843 | 146 | |
BC016422.1 | - | 8 - 9 | 806 | 3964935 | 146 | |
M65029.1 | - | 8 - 9 | 795 | 146 | ||
M93567.1 | - | 8 - 9 | 785 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |