Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024184.mm.1 |
NM_001033543.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024184.mm.1 |
NM_001033543.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024184.mm.1 |
TC0900002934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024186.mm.1 |
NM_001033543.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024186.mm.1 |
NM_001033543.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024186.mm.1 |
TC0900002934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024188.mm.1 |
NM_001033543.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024188.mm.1 |
NM_001033543.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024188.mm.1 |
TC0900002934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024191.mm.1 |
NM_001033543.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024191.mm.1 |
NM_001033543.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024191.mm.1 |
TC0900002934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024192.mm.1 |
NM_001033543.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024192.mm.1 |
NM_001033543.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024192.mm.1 |
TC0900002934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024189.mm.1 |
NM_001033543.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024189.mm.1 |
NM_001033543.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024189.mm.1 |
TC0900002934.mm.1 |
MTA TC |