Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253831.1 | NP_001240760.1 | 20 - 21 | 2094 | 63 | |
NM_001253834.1 | NP_001240763.1 | 21 - 22 | 2185 | 63 | ||
NM_001253836.1 | NP_001240765.1 | 20 - 21 | 2117 | 63 | ||
NM_175025.4 | NP_778190.3 | 21 - 22 | 2300 | 63 | ||
GenBank mRNA | AJ551270.1 | - | 19 - 20 | 1882 | 63 | |
AK028393.1 | - | 20 - 21 | 2063 | 63 | ||
AK133703.1 | - | 20 - 21 | 2095 | 63 | ||
AK159918.1 | - | 21 - 22 | 2251 | 63 | ||
AK161432.1 | - | 5 - 6 | 505 | 63 | ||
AK163049.1 | - | 21 - 22 | 2146 | 63 | ||
AK173154.1 | - | 19 - 20 | 1873 | 63 | ||
BC043091.1 | - | 21 - 22 | 2266 | 6405251 | 63 | |
BC043706.1 | - | 21 - 22 | 2246 | 63 | ||
BU754887.1 | - | 1 - 2 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |