Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022328.2 | NP_071723.1 | 4 - 5 | 530 | 53 | |
GenBank mRNA | AF298887.1 | - | 3 - 4 | 416 | 53 | |
AF312858.1 | - | 4 - 5 | 427 | 53 | ||
AK144605.1 | - | 4 - 5 | 558 | 53 | ||
AK147804.1 | - | 1 - 2 | 135 | 53 | ||
AK156820.1 | - | 4 - 5 | 557 | 53 | ||
BC053005.1 | - | 4 - 5 | 530 | 5716396 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |