Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133193.3 | NP_573456.1 | 4 - 5 | 695 | 113 | |
GenBank mRNA | AF284433.1 | - | 4 - 5 | 700 | 113 | |
AK085780.1 | - | 4 - 5 | 563 | 113 | ||
BC117797.2 | - | 4 - 5 | 696 | 40092208 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |