Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081071.2 | NP_001074540.1 | 2 - 3 | 368 | 71 | |
NM_001177967.1 | NP_001171438.1 | 2 - 3 | 347 | 71 | ||
NM_001177968.1 | NP_001171439.1 | 2 - 3 | 336 | 71 | ||
GenBank mRNA | AK144580.1 | - | 2 - 3 | 311 | 71 | |
BC147499.1 | - | 2 - 3 | 286 | 9056361 | 71 | |
BC150884.1 | - | 2 - 3 | 275 | 9087795 | 71 | |
BC158076.1 | - | 2 - 3 | 307 | 9088168 | 71 | |
BC158079.1 | - | 2 - 3 | 307 | 9088171 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |