Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033128.3 | NP_001028300.1 | 2 - 3 | 146 | 58 | |
GenBank mRNA | AK037753.1 | - | 2 - 3 | 132 | 58 | |
AK045850.1 | - | 2 - 3 | 143 | 58 | ||
AK161343.1 | - | 2 - 3 | 140 | 58 | ||
AK168901.1 | - | 2 - 3 | 147 | 58 | ||
BC076577.1 | - | 2 - 3 | 135 | 58 | ||
BY063920.1 | - | 2 - 3 | 147 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |