Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081337.1 | NP_001074806.1 | 12 - 13 | 3751 | 98 | |
GenBank mRNA | AK028440.1 | - | 12 - 13 | 2865 | 98 | |
AK122501.1 | - | 10 - 11 | 2028 | 98 | ||
BC058408.1 | - | 3 - 4 | 224 | 98 | ||
BC072593.1 | - | 14 - 15 | 4037 | 98 | ||
BC089367.1 | - | 2 - 3 | 137 | 98 | ||
BC150682.1 | - | 12 - 13 | 3796 | 9087593 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |