Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122594.2 | NP_001116066.2 | 6 - 7 | 1000 | 79 | |
GenBank mRNA | AK048260.1 | - | 6 - 7 | 957 | 79 | |
AK048472.1 | - | 6 - 7 | 1001 | 79 | ||
BC117961.1 | - | 6 - 7 | 1000 | 40098281 | 79 | |
BC117962.1 | - | 6 - 7 | 992 | 40098282 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |