Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145823.2 | NP_665822.1 | 3 - 4 | 1064 | 65 | |
GenBank mRNA | AB077281.1 | - | 3 - 4 | 1045 | 65 | |
AB077282.1 | - | 3 - 4 | 342 | 65 | ||
AK003844.1 | - | 2 - 3 | 312 | 65 | ||
AK021206.1 | - | 3 - 4 | 277 | 65 | ||
AK153625.1 | - | 3 - 4 | 1065 | 65 | ||
AK171340.1 | - | 3 - 4 | 1091 | 65 | ||
AK171918.1 | - | 3 - 4 | 1061 | 65 | ||
AK172137.1 | - | 3 - 4 | 1065 | 65 | ||
BC082333.1 | - | 3 - 4 | 896 | 65 | ||
BC108351.1 | - | 3 - 4 | 924 | 6312193 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |