Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028877.1 | NP_083153.1 | 3 - 4 | 189 | 73 | |
GenBank mRNA | AK014493.1 | - | 3 - 4 | 193 | 73 | |
BC113196.1 | - | 3 - 4 | 192 | 73 | ||
BC113802.1 | - | 3 - 4 | 187 | 73 | ||
EF208034.1 | - | 3 - 4 | 189 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |