Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133660.3 | NP_598421.1 | 6 - 7 | 993 | 81 | |
GenBank mRNA | AK144549.1 | - | 6 - 7 | 978 | 81 | |
BC019208.1 | - | 6 - 7 | 924 | 5052893 | 81 | |
S80191.1 | - | 6 - 7 | 923 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |