Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21756067 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21756066 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21756069 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2063366 |
BC065794.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2063366 |
BC092215.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2063366 |
BC094216.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2063366 |
AK169037.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2063366 |
AK212797.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2178743 |
AK046582.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
261633 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312974 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556082 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
321523 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
739058 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AF525925.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AK049061.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AK077496.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AK089289.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AK129052.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AK138637.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AK161677.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
AK170739.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
BC048948.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
BC056952.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
BC129804.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
NM_001039200.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849258 |
NM_146157.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
610636 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351628 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
150202 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P149396 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P176335 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965592 |
AV146140.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965592 |
AK078774.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965592 |
NM_001195390.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965592 |
NM_133850.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2063366 |
BC065794.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2063366 |
BC092215.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2063366 |
BC094216.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2063366 |
AK169037.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2063366 |
AK212797.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2178743 |
AK046582.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756056 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756063 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756065 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756066 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756080 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756078 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756058 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756081 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756069 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756075 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756071 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756067 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756061 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756060 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21756074 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04744930 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04744905 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P149396 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P646705 |
BC065794.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P646705 |
BC092215.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P646705 |
BC094216.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P646705 |
AK169037.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P646705 |
BC003310.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115521 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P149396 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01353857 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01582468 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01785799 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01353861 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01243667 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01243668 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21756069 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21756066 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21756067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21756035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21755958 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01928828 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755962 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755961 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756034 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756088 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755955 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02154460 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756052 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756060 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756092 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756032 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755975 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756055 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756054 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756071 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756064 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756033 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756049 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755973 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756031 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756038 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756081 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756070 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02207530 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755956 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02267411 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756076 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755968 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756085 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01243673 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01991910 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01941823 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01615370 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01353861 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756069 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01958360 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755969 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756056 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756080 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01638025 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756082 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756063 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756090 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755965 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01945276 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01301716 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755967 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756059 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756075 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756074 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755963 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756047 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21756045 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21755960 |
- |
- |