Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144831.2 | NP_659080.2 | 5 - 6 | 592 | 69 | |
GenBank mRNA | AK078552.1 | - | 5 - 6 | 531 | 69 | |
AK142226.1 | - | 5 - 6 | 526 | 69 | ||
AK170616.1 | - | 5 - 6 | 520 | 69 | ||
AK172600.1 | - | 5 - 6 | 532 | 69 | ||
AK220485.1 | - | 5 - 6 | 551 | 69 | ||
BC065064.1 | - | 5 - 6 | 521 | 69 | ||
BC132445.1 | - | 5 - 6 | 576 | 40130722 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |