Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177388.3 | NP_796362.1 | 3 - 4 | 901 | 92 | |
GenBank mRNA | AK038549.1 | - | 3 - 4 | 902 | 92 | |
BC138146.1 | - | 2 - 3 | 739 | 8861165 | 92 | |
BC138147.1 | - | 2 - 3 | 739 | 8861166 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |