Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028524.3 | NP_082800.1 | 2 - 3 | 430 | 68 | |
GenBank mRNA | AK005970.1 | - | 2 - 3 | 424 | 68 | |
AY162282.1 | - | 2 - 3 | 310 | 68 | ||
AY241872.1 | - | 2 - 3 | 424 | 68 | ||
BC050983.1 | - | 2 - 3 | 427 | 68 | ||
BC060997.1 | - | 2 - 3 | 323 | 6742828 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |