Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Llgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252532.1 | NP_001239461.1 | 22 - 23 | 3148 | 63 | |
NM_145438.2 | NP_663413.2 | 22 - 23 | 3076 | 63 | ||
GenBank mRNA | AK078901.1 | - | 6 - 7 | 807 | 63 | |
AK143486.1 | - | 22 - 23 | 3137 | 63 | ||
AK143798.1 | - | 22 - 23 | 3053 | 63 | ||
AK167536.1 | - | 22 - 23 | 3048 | 63 | ||
AY033650.1 | - | 22 - 23 | 3026 | 63 | ||
BC013232.1 | - | 18 - 19 | 2617 | 63 | ||
BC037226.1 | - | 22 - 23 | 3048 | 3584689 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |