Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037726.1 | NP_001032815.1 | 5 - 6 | 793 | 66 | |
NM_009952.2 | NP_034082.1 | 7 - 8 | 957 | 66 | ||
NM_133828.2 | NP_598589.2 | 6 - 7 | 915 | 66 | ||
GenBank mRNA | AF448507.1 | - | 7 - 8 | 833 | 66 | |
AF448508.1 | - | 1485 | 66 | |||
AK044992.1 | - | 6 - 7 | 730 | 66 | ||
AK044993.1 | - | 6 - 7 | 730 | 66 | ||
AK079223.1 | - | 6 - 7 | 911 | 66 | ||
BB831364.1 | - | 2 - 3 | 287 | 66 | ||
BC021649.1 | - | 6 - 7 | 848 | 4017170 | 66 | |
X92497.1 | - | 5 - 6 | 770 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |