Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023631.2 | NP_076120.2 | 30 - 31 | 3602 | 111 | |
GenBank mRNA | AF233581.1 | - | 30 - 31 | 3529 | 111 | |
AK009112.1 | - | 5 - 6 | 521 | 111 | ||
AK158427.1 | - | 30 - 31 | 3603 | 111 | ||
BC117975.1 | - | 30 - 31 | 3529 | 40098789 | 111 | |
BC117976.1 | - | 30 - 31 | 3529 | 40098791 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |