Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011729.2 | NP_035859.2 | 6 - 7 | 988 | 65 | |
GenBank mRNA | AK137563.1 | - | 6 - 7 | 820 | 65 | |
AK140825.1 | - | 6 - 7 | 817 | 65 | ||
AK160645.1 | - | 4 - 5 | 360 | 65 | ||
AK164205.1 | - | 6 - 7 | 806 | 65 | ||
BC033413.1 | - | 6 - 7 | 708 | 65 | ||
BC053034.1 | - | 6 - 7 | 797 | 65 | ||
BC060659.1 | - | 6 - 7 | 787 | 65 | ||
BC092235.1 | - | 6 - 7 | 779 | 65 | ||
BC141070.1 | - | 6 - 7 | 818 | 9055682 | 65 | |
D16306.1 | - | 6 - 7 | 683 | 65 | ||
U40796.1 | - | 6 - 7 | 884 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |