Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146104.3 | NP_666216.1 | 2 - 3 | 535 | 105 | |
NM_146134.2 | NP_666246.1 | 2 - 3 | 535 | 105 | ||
GenBank mRNA | AK050828.1 | - | 1 - 2 | 528 | 105 | |
AK148400.1 | - | 2 - 3 | 488 | 105 | ||
AK152334.1 | - | 2 - 3 | 478 | 105 | ||
BC012406.1 | - | 2 - 3 | 460 | 3483214 | 105 | |
BC024111.1 | - | 2 - 3 | 401 | 5098973 | 105 | |
BC057865.1 | - | 2 - 3 | 478 | 5320175 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |