Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111320.1 | NP_001104790.1 | 2 - 3 | 307 | 119 | |
NM_010497.3 | NP_034627.3 | 2 - 3 | 285 | 119 | ||
GenBank mRNA | AF020039.1 | - | 1 - 2 | 135 | 119 | |
AK087063.1 | - | 2 - 3 | 187 | 119 | ||
AK149019.1 | - | 2 - 3 | 216 | 119 | ||
AK151212.1 | - | 2 - 3 | 210 | 119 | ||
AK159173.1 | - | 2 - 3 | 216 | 119 | ||
AK160896.1 | - | 2 - 3 | 217 | 119 | ||
AK167158.1 | - | 2 - 3 | 257 | 119 | ||
AK167583.1 | - | 2 - 3 | 221 | 119 | ||
BC088986.1 | - | 2 - 3 | 257 | 6808731 | 119 | |
CF950262.1 | - | 2 - 3 | 246 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |