Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139270.2 | NP_644676.1 | 8 - 9 | 1244 | 93 | |
GenBank mRNA | AF132083.1 | - | 2 - 3 | 164 | 93 | |
AK045576.1 | - | 8 - 9 | 1244 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Mus musculus Inventoried Probe spans exons |