Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK047751.1 | - | 4 - 5 | 939 | 80 | |
AK135543.1 | - | 4 - 5 | 947 | 80 | ||
AK156828.1 | - | 4 - 5 | 944 | 80 | ||
BC024418.1 | - | 4 - 5 | 982 | 4982610 | 80 | |
BC026567.1 | - | 4 - 5 | 970 | 4982239 | 80 | |
BC085273.1 | - | 4 - 5 | 922 | 30144329 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |