Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194355.2 | NP_919336.1 | 9 - 10 | 1140 | 77 | |
GenBank mRNA | BC046486.1 | - | 2 - 3 | 223 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |