Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178928.4 | NP_849259.2 | 7 - 8 | 819 | 66 | |
GenBank mRNA | AK035227.1 | - | 7 - 8 | 819 | 66 | |
AK053714.1 | - | 7 - 8 | 818 | 66 | ||
BC138923.1 | - | 7 - 8 | 780 | 8861945 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |