Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159344.1 | NP_001152816.1 | 9 - 10 | 2018 | 102 | |
NM_027195.2 | NP_081471.2 | 9 - 10 | 2018 | 102 | ||
GenBank mRNA | AK138981.1 | - | 4 - 5 | 805 | 102 | |
AK173341.1 | - | 4 - 5 | 808 | 102 | ||
BC075673.1 | - | 9 - 10 | 2020 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |