Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146089.2 | NP_666201.1 | 7 | 734 | 105 | |
GenBank mRNA | AK076912.1 | - | 7 | 714 | 105 | |
AK146923.1 | - | 7 | 731 | 105 | ||
BC024400.1 | - | 7 | 734 | 5026575 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Amplicon spans exons and probe does not span exons |