Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037717.3 | NP_001032806.2 | 7 - 8 | 644 | 97 | |
GenBank mRNA | BC157917.1 | - | 7 - 8 | 593 | 9007345 | 97 |
BC157928.1 | - | 7 - 8 | 593 | 9088101 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |