Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252614.1 | NP_001239543.1 | 11 - 12 | 1859 | 132 | |
NM_001252615.1 | NP_001239544.1 | 5 - 6 | 933 | 132 | ||
NM_001252616.1 | NP_001239545.1 | 4 - 5 | 953 | 132 | ||
NM_016893.5 | NP_058589.2 | 10 - 11 | 1770 | 132 | ||
NR_045554.1 | 10 - 11 | 2189 | 132 | |||
GenBank mRNA | AK048520.1 | - | 10 - 11 | 1908 | 132 | |
AK051811.1 | - | 10 - 11 | 1727 | 132 | ||
AK134537.1 | - | 10 - 11 | 1863 | 132 | ||
BC010666.1 | - | 11 - 12 | 1838 | 3594582 | 132 | |
EF591874.1 | - | 5 - 6 | 933 | 132 | ||
EF591875.1 | - | 4 - 5 | 953 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |