Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113559.2 | NP_001107031.2 | 9 - 10 | 1393 | 75 | |
NM_001243163.1 | NP_001230092.1 | 9 - 10 | 1285 | 75 | ||
NM_011444.3 | NP_035574.2 | 10 - 11 | 1537 | 75 | ||
GenBank mRNA | AB006330.1 | - | 10 - 11 | 1449 | 75 | |
AJ010604.1 | - | 9 - 10 | 1275 | 75 | ||
AK015212.1 | - | 3 - 4 | 346 | 75 | ||
AK029047.1 | - | 12 - 13 | 1535 | 75 | ||
AK160154.1 | - | 3 - 4 | 351 | 75 | ||
BC110478.2 | - | 9 - 10 | 1277 | 40047865 | 75 | |
KF793916.1 | - | 10 - 11 | 1238 | 75 | ||
X65657.1 | - | 3 - 4 | 322 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |