Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133879.2 | NP_598640.2 | 6 - 7 | 1336 | 55 | |
GenBank mRNA | AK027912.1 | - | 5 - 6 | 1820 | 55 | |
AM161454.1 | - | 6 - 7 | 1284 | 55 | ||
CX202255.1 | - | 4 - 5 | 513 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |