Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028307.1 | NP_082583.1 | 7 - 8 | 1122 | 89 | |
GenBank mRNA | BC049363.1 | - | 7 - 8 | 1122 | 6490411 | 89 |
BC057030.1 | - | 7 - 8 | 1138 | 6833743 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |