Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081073.1 | NP_001074542.1 | 9 - 10 | 1451 | 66 | |
GenBank mRNA | AK035879.1 | - | 8 - 9 | 1231 | 66 | |
BC119216.1 | - | 9 - 10 | 1332 | 8733965 | 66 | |
BC119218.1 | - | 9 - 10 | 1332 | 8733967 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |