Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285931.1 | NP_001272860.1 | 6 - 7 | 2389 | 119 | |
NM_001285932.1 | NP_001272861.1 | 6 - 7 | 2155 | 119 | ||
NM_027883.3 | NP_082159.3 | 6 - 7 | 1880 | 119 | ||
GenBank mRNA | AK004733.1 | - | 6 - 7 | 1881 | 119 | |
AK036763.1 | - | 2 - 3 | 281 | 119 | ||
AK076217.1 | - | 5 - 6 | 1856 | 119 | ||
AK129062.1 | - | 6 - 7 | 1863 | 119 | ||
BC058370.1 | - | 6 - 7 | 2217 | 119 | ||
BC080856.1 | - | 6 - 7 | 2155 | 6854931 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |