Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009498.4 | NP_033524.1 | 2 - 3 | 163 | 88 | |
GenBank mRNA | AK051272.1 | - | 2 - 3 | 164 | 88 | |
AK051691.1 | - | 2 - 3 | 158 | 88 | ||
AK168413.1 | - | 2 - 3 | 152 | 88 | ||
AK169056.1 | - | 2 - 3 | 161 | 88 | ||
AK169087.1 | - | 2 - 3 | 155 | 88 | ||
BC060045.1 | - | 2 - 3 | 138 | 5690213 | 88 | |
U60961.1 | - | 2 - 3 | 105 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |