Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Th | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009377.1 | NP_033403.1 | 2 - 3 | 315 | 66 | |
GenBank mRNA | M69200.1 | - | 2 - 3 | 315 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |