Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082960.1 | NP_001076429.1 | 7 - 8 | 804 | 79 | |
NM_008401.2 | NP_032427.2 | 7 - 8 | 804 | 79 | ||
GenBank mRNA | AK154224.1 | - | 7 - 8 | 724 | 79 | |
AK154474.1 | - | 7 - 8 | 758 | 79 | ||
AK154798.1 | - | 7 - 8 | 720 | 79 | ||
AK155520.1 | - | 7 - 8 | 729 | 79 | ||
AK155671.1 | - | 7 - 8 | 770 | 79 | ||
AK155717.1 | - | 7 - 8 | 762 | 79 | ||
AK170324.1 | - | 7 - 8 | 776 | 79 | ||
AK171068.1 | - | 4 - 5 | 433 | 79 | ||
AK171150.1 | - | 7 - 8 | 739 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |