Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009135.2 | NP_033161.2 | 9 - 10 | 1295 | 127 | |
GenBank mRNA | AK142964.1 | - | 9 - 10 | 1300 | 127 | |
BC145273.1 | - | 9 - 10 | 1152 | 127 | ||
L36179.1 | - | 9 - 10 | 1331 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |