Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008653.2 | NP_032679.2 | 2 - 3 | 343 | 76 | |
GenBank mRNA | AF097333.1 | - | 2 - 3 | 294 | 76 | |
AK146661.1 | - | 2 - 3 | 344 | 76 | ||
AK146886.1 | - | 2 - 3 | 344 | 76 | ||
AK169302.1 | - | 2 - 3 | 324 | 76 | ||
AK169306.1 | - | 2 - 3 | 324 | 76 | ||
BC131672.1 | - | 2 - 3 | 319 | 76 | ||
BC154408.1 | - | 2 - 3 | 364 | 40110890 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |