Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177331.4 | NP_796305.3 | 12 - 13 | 1485 | 86 | |
GenBank mRNA | AK030995.1 | - | 12 - 13 | 1495 | 86 | |
AK031077.1 | - | 12 - 13 | 1464 | 86 | ||
AK158255.1 | - | 11 - 12 | 1308 | 86 | ||
BC090653.1 | - | 12 - 13 | 1485 | 30659763 | 86 | |
BC138157.1 | - | 12 - 13 | 1372 | 8861176 | 86 | |
BC138158.1 | - | 12 - 13 | 1372 | 8861177 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |