Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029338.3 | NP_083614.1 | 3 - 4 | 574 | 79 | |
GenBank mRNA | AK006440.1 | - | 3 - 4 | 577 | 79 | |
AK140720.1 | - | 1 - 2 | 55 | 79 | ||
AK165820.1 | - | 3 - 4 | 445 | 79 | ||
BC019423.1 | - | 3 - 4 | 548 | 4922914 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |